Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CBLN1 Mouse Monoclonal Antibody [clone: Rad51D 5A8/4]

Supplier: Genetex

GTX11051
89328-216EA 358.96 USD InStock
89328-216
Anti-CBLN1 Mouse Monoclonal Antibody [clone: Rad51D 5A8/4]
Antibodies
Mouse monoclonal [Rad51D 5A8 / 4] to Rad51D

WB: Use at an assay dependent dilution. Predicted molecular weight: 39 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CBLN1
Clonality: Monoclonal
Clone: Rad51D 5A8/4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty