Anti-INSL4 Mouse Monoclonal Antibody [clone: M148]

Supplier: GeneTex
GTX11024
89328-206EA 630.11 USD
89328-206
Anti-INSL4 Mouse Monoclonal Antibody [clone: M148]
Antibodies
Mouse monoclonal antibody [M148] to CD41

IP: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: INSL4
Clonality: Monoclonal
Clone: M148
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR