Anti-TMEM30C Rabbit Polyclonal Antibody

GTX110174
89311-050EA 176.03 USD
89311-050
Anti-TMEM30C Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to MESP2 (mesoderm posterior 2 homolog (mouse))

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: TMEM30C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR