Anti-PMP22 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX110098
89320-462EA 582.76 USD
89320-462
Anti-PMP22 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RBMY1A1 (RNA binding motif protein, Y-linked, family 1, member A1)

  • Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PMP22
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR