Anti-OR10Z1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX110079
89330-472EA 582.76 USD
89330-472
Anti-OR10Z1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to HSPA2 (heat shock-related 70 kDa protein 2)

Suggested starting dilutions are as follows: WB: 1:5000-1:20000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OR10Z1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR