Anti-NEFH Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX110065
89320-454EA 582.76 USD
89320-454
Anti-NEFH Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to 200 kD Neurofilament Heavy (neurofilament, heavy polypeptide 200kDa)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-Fr: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NEFH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR