Anti-N4BP1 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX110034
89320-452EA 427.1 USD
89320-452
Anti-N4BP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Villin (villin 1)

Suggested starting dilutions are as follows: WB: 1:5000-1:20000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: N4BP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR