Anti-hnRNP G Rabbit Polyclonal Antibody

GTX109925
89310-970EA 176.03 USD
89310-970
Anti-hnRNP G Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to TLE1 (transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila))

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: hnRNP G
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR