Anti-OLFML2A Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX109917
89335-458EA 582.76 USD
89335-458
Anti-OLFML2A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to BAAT (bile acid CoA: amino acid N-acyltransferase (glycine N-choloyltransferase))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OLFML2A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR