Anti-PABPC5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX109885
89330-470EA 582.76 USD
89330-470
Anti-PABPC5 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TCF3 (transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47))

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PABPC5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR