Anti-ENDOGL1 Rabbit Polyclonal Antibody

GTX109873
89310-960EA 176.03 USD
89310-960
Anti-ENDOGL1 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Topo III beta-1 (topoisomerase (DNA) III beta)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: ENDOGL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR