Anti-OR51D1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX109827
89320-420EA 434.85 USD
89320-420
Anti-OR51D1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SMC3 (structural maintenance of chromosomes 3)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OR51D1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR