Anti-UBAP2L Goat Polyclonal Antibody

Supplier: GeneTex
GTX10979
89280-420EA 557.17 USD
89280-420
Anti-UBAP2L Goat Polyclonal Antibody
Antibodies
Goat polyclonal to Leptin Receptor

ELISA: Use at a concentration of 0.5 - 1 ?g/ml. This concentration detects a limit of 0.12 ng/well of mouse Leptin receptor. WB: Use at a concentration of 0.1 - 0.2 ?g/ml. This concentration was determined for mouse Leptin receptor using 5 ng/lane under non-reducing and reducing conditions. Predicted molecular weight: 81 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: UBAP2L
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR