Anti-OR6C70 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX109699
89320-394EA 582.76 USD
89320-394
Anti-OR6C70 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to APBB3 (amyloid beta (A4) precursor protein-binding, family B, member 3)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OR6C70
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR