Anti-GHITM Rabbit Polyclonal Antibody

Supplier: Genetex

GTX109689
89320-392EA 394.91 USD
89320-392
Anti-GHITM Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PKC theta-phospho-S695 (protein kinase C, theta)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PRKCQ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR