Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SCGB2A1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX109674
89310-892EA 345 USD InStock
89310-892
Anti-SCGB2A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to FURIN (furin (paired basic amino acid cleaving enzyme))

Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: SCGB2A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty