Anti-LDLRAD3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX109620
89320-376EA 642.92 USD
89320-376
Anti-LDLRAD3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to p53R2 (ribonucleotide reductase M2 B (TP53 inducible))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: LDLRAD3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR