Anti-SLC30A1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX109585
89320-368EA 582.76 USD
89320-368
Anti-SLC30A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Troponin T1 (slow skeletal) (troponin T type 1 (skeletal, slow))

  • Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SLC30A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR