Anti-OR5B12 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX109576
89310-852EA 837.06 USD
89310-852
Anti-OR5B12 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TIN2 (TERF1 (TRF1)-interacting nuclear factor 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OR5B12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR