Anti-OR51B2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX109573
89310-850EA 582.76 USD
89310-850
Anti-OR51B2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ubiquitin B (ubiquitin B)

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OR51B2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR