Anti-ACBD6 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX109566
89320-356EA 394.91 USD
89320-356
Anti-ACBD6 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Proteasome 20S beta 3 (proteasome (prosome, macropain) subunit, beta type, 3)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ACBD6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR