Anti-STEA2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX109559
89320-348EA 582.76 USD
89320-348
Anti-STEA2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to VAX1 (ventral anterior homeobox 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: STEA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR