Anti-F13B Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX109533
89310-834EA 837.06 USD
89310-834
Anti-F13B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PDP 1 (protein phosphatase 2C, magnesium-dependent, catalytic subunit)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: F13B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR