Anti-EIF2B1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX109525
89320-344EA 394.91 USD
89320-344
Anti-EIF2B1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to EIF2B1 (eukaryotic translation initiation factor 2B, subunit 1 alpha, 26kDa)

Suggested starting dilutions are as follows: WB: 1:5000-1:20000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: EIF2B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR