Anti-YEATS2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX109433
89310-820EA 176.03 USD
89310-820
Anti-YEATS2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ETFDH (electron-transferring-flavoprotein dehydrogenase)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: YEATS2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR