Anti-GFM2 Rabbit Polyclonal Antibody

GTX109295
89310-784EA 176.03 USD
89310-784
Anti-GFM2 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to PIR1 (dual specificity phosphatase 11 (RNA/RNP complex 1-interacting))

Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: DUSP11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR