Anti-HMG14 Goat Polyclonal Antibody

Supplier: GeneTex
GTX10925
89287-852EA 607.87 USD
89287-852
Anti-HMG14 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to IRF6

Approx 60kDa band observed in Human Ovary lysates (calculated MW of 53.1kDa according to NP_006138.1). In transfected HEK293 transiently expressing IRF6 a band of approx. 55kDa is observed. This band is not observed in the non-transfected HEK293. Recommended for use at 0.2-0.6µg/ml. In paraffin embedded Human Kidney shows strong nuclear staining in DCT and glomerulus. Recommended concentration, 2-3µg/ml.

Type: Primary
Antigen: HMG14
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR