Anti-BRWD3 Goat Polyclonal Antibody

Supplier: GeneTex
GTX10924
89261-508EA 614.99 USD
89261-508
Anti-BRWD3 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to BCAP31 / BAP31

Approx 30kDa band observed in Human Placenta and Brain lysate (predicted MW of 28kDa according to NP_005736). Recommended for use at 1-3µg/ml. This antibody is expected to recognise isoform a (NP_001132929.1) and isoform b (NP_005736.3; NP_001132913.1).

Type: Primary
Antigen: BRWD3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR