Anti-CADM3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX109207
89320-314EA 582.56 USD
89320-314
Anti-CADM3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Beta crystallin A3 (crystallin, beta A1)

Suggested starting dilutions are as follows: WB: 1:5000-1:20000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CADM3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR