Anti-ARMC6 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX109096
89320-288EA 394.91 USD
89320-288
Anti-ARMC6 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to HMGCL (3-hydroxymethyl-3-methylglutaryl-CoA lyase)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ARMC6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR