Anti-hnRNP H Mouse Monoclonal Antibody [clone: TRAP1.3.6]

Supplier: Genetex

GTX10909
89286-774EA 425.13 USD
89286-774
Anti-hnRNP H Mouse Monoclonal Antibody [clone: TRAP1.3.6]
Antibodies
Mouse monoclonal antibody [TRAP-1] to CD40L

Recommended Starting Dilutions: Flow Cytometry: 1/10 - 1/50. Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: hnRNP H
Clonality: Monoclonal
Clone: TRAP1.3.6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR