Anti-OR8H3 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX108921
89320-258EA 394.91 USD
89320-258
Anti-OR8H3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to LOC169436 (chromosome 9 open reading frame 96)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OR8H3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR