Anti-SERPINA11 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX108871
89310-652EA 1199.03 USD
89310-652
Anti-SERPINA11 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CYP7A1 (cytochrome P450, family 7, subfamily A, polypeptide 1)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: SERPINA11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR