Anti-OR10J3 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX108819
89320-244EA 427.1 USD
89320-244
Anti-OR10J3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to LILRA3 (leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member 3)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OR10J3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR