Anti-STK36 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX108794
89320-240EA 642.92 USD
89320-240
Anti-STK36 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to STK36 (serine/threonine kinase 36, fused homolog (Drosophila))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: STK36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR