Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

Anti-OR10C1 Mouse Monoclonal Antibody [clone: 6F11/2]

Supplier: Genetex

GTX10873
89317-248EA 315.66 USD InStock
89317-248
Anti-OR10C1 Mouse Monoclonal Antibody [clone: 6F11/2]
Antibodies
Type: Primary
Antigen: OR10C1
Clonality: Monoclonal
Clone: 6F11/2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty