Anti-HEY2 Mouse Monoclonal Antibody [clone: GA6]

Supplier: Genetex

GTX10870
89317-246EA 405.23 USD
89317-246
Anti-HEY2 Mouse Monoclonal Antibody [clone: GA6]
Antibodies
Mouse monoclonal antibody [GA6] to Granzyme A

Recommended Starting Dilutions: Immunohistology - Paraffin: 1/50 - 1/200. Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: HEY2
Clonality: Monoclonal
Clone: GA6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR