Anti-BRI3B Rabbit Polyclonal Antibody

Supplier: Genetex

GTX108658
89320-214EA 394.91 USD
89320-214
Anti-BRI3B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to FAM18B (family with sequence similarity 18, member B)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: BRI3B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR