Anti-MRPS33 Rabbit Polyclonal Antibody

GTX108643
89310-590EA 176.03 USD
89310-590
Anti-MRPS33 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Tango13

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: MRPS33
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR