Anti-G3BP2 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX108630
89310-582EA 394.91 USD
89310-582
Anti-G3BP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: STAT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR