Anti-RBM6 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX108588
89320-190EA 582.76 USD
89320-190
Anti-RBM6 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Ras GRF2 (Ras protein-specific guanine nucleotide-releasing factor 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RBM6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR