Anti-LATS1/2 Rabbit Polyclonal Antibody

GTX108518
89310-528EA 176.03 USD
89310-528
Anti-LATS1/2 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to PGRP-S (peptidoglycan recognition protein 1)

Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: LATS1/2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR