Anti-PECI Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX108500
89320-170EA 427.1 USD
89320-170
Anti-PECI Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to NKG2-C (killer cell lectin-like receptor subfamily C, member 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PECI
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR