Anti-MAP3K10 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX108479
89316-536EA 395.16 USD
89316-536
Anti-MAP3K10 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CD3 zeta(CD247) (CD3-zeta(CD247) molecule)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MAP3K10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR