Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

Anti-STEAP4 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX108461
89310-486EA 167.65 USD InStock
89310-486
Anti-STEAP4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CXCL5

Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: STEAP4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty