Anti-CASP6 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX108423
89320-142EA 427.1 USD
89320-142
Anti-CASP6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Interferon alpha-6 (interferon, alpha 6)

Suggested starting dilutions are as follows: WB: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CASP6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR