Anti-CATD Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX108421
89316-528EA 727.73 USD
89316-528
Anti-CATD Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to IL-1F5 (interleukin 1 family, member 5 (delta))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CATD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR