Anti-EFNA2 Mouse Monoclonal Antibody [clone: 17001.31]

Supplier: GeneTex
GTX10842
89316-016EA 623.55 USD
89316-016
Anti-EFNA2 Mouse Monoclonal Antibody [clone: 17001.31]
Antibodies
Mouse monoclonal [17001.31] to Oncostatin M

ELISA: Use at a concentration of 0.5-1 ?g/ml. Neut: ND50 = 0.1 - 5 ?g/mlWB: Use at a concentration of 1-2 ?g/ml. Predicted molecular weight: 22 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: EFNA2
Clonality: Monoclonal
Clone: 17001.31
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR