Anti-ITGA6 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX108371
89310-378EA 793.5 USD
89310-378
Anti-ITGA6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ABCA4 (ATP-binding cassette, sub-family A (ABC1), member 4)

Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: ITGA6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR