Anti-GPR21 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX108303
89320-106EA 394.91 USD
89320-106
Anti-GPR21 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to COL4A3BP(collagen, type IV, alpha 3 (Goodpasture antigen) binding protein)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: COL4A3BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR